Learn More
MOLECULAR DEPOT LLC Human Pooled Serum - 50 mL
Supplier:  MOLECULAR DEPOT LLC S2010007
Encompass_Preferred
Normal Human Pooled Serum (50 mL) consists of pooled human normal serum obtained from normal human clotted blood Serum is not filtered and contains no additives or preservatives It is supplied as a yellow-orange liquid For in vitro research use only
Catalog No. 50-000-13456