Learn More
Active Motif Human Positive Control Primer Set MYT1
SDP
Catalog No. NC1850708
Encompass
Human Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with human samples. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP samples from multiple human cell lines.
Catalog No. NC1850708 Supplier Active Motif Supplier No. 71007
Human Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with human samples. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP samples from multiple human cell lines.