Learn More
BIOHIPPO Human POSTN Microsample
SDP
Catalog No. 5000315531
Encompass
Human POSTN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of POSTN in serum plasma and other biological fluids It provides a detection range of 7 82-500 ng/mL with a sensitivity of 2 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human POSTN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human POSTN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human POSTN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15531 Supplier BIOHIPPO Supplier No. BHE15212566
Human POSTN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of POSTN in serum plasma and other biological fluids It provides a detection range of 7 82-500 ng/mL with a sensitivity of 2 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human POSTN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human POSTN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human POSTN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color