Learn More
Biomatik Corporation Human Potassium Inwardly Rectifying Channel Subfamily J, Member 10 (KCNJ10) ELISA Kit
Supplier:  Biomatik Corporation EKU08489
Encompass_Preferred
Human Potassium Inwardly Rectifying Channel Subfamily J, Member 10 (KCNJ10) ELISA Kit for detecting KCNJ10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P78508. Gene ID: 3766. Target Synonyms: Kir4.1; Kir1.2; ATP-sensitive inward rectifier potassium channel 10; ATP-dependent inwardly rectifying potassium channel Kir4.1; Inward rectifier K (+) channel Kir1.2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4926