Learn More
Biomatik Corporation Human POTE Ankyrin Domain Family, Member G (POTEG) CLIA Kit
Supplier:  Biomatik Corporation EKU09493
Encompass_Preferred
Human POTE Ankyrin Domain Family, Member G (POTEG) CLIA Kit for detecting POTEG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q6S5H5. Gene ID: 404785. Target Synonyms: A26C2; POTE14; POTE14a; CT104.4; ANKRD26-Like Family C Member 2; Cancer/Testis Antigen Family 104,Member 4; Prostate, ovary, testis-expressed protein on chromosome 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6682