Learn More
BIOHIPPO Human POTEE Microsample
SDP
Catalog No. 5000314406
Encompass
Human POTEE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of POTEE in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 15 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human POTEE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human POTEE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human POTEE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14406 Supplier BIOHIPPO Supplier No. BHE15212390
Human POTEE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of POTEE in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 15 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human POTEE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human POTEE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human POTEE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color