Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human POU5F1B (aa 51-101) Control Fragment Recombinant Protein

Catalog No. RP105098
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105098 Supplier Invitrogen™ Supplier No. RP105098
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66155. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This intronless gene was thought to be a transcribed pseudogene of POU class 5 homeobox 1, however, it has been reported that this gene can encode a functional protein. The encoded protein is nearly the same length as and highly similar to the POU class 5 homeobox 1 transcription factor, has been shown to be a weak transcriptional activator and may play a role in carcinogenesis and eye development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q06416
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 5462
Name Human POU5F1B (aa 51-101) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias OCT4PG1; Oct4-pg1; octamer binding protein 3_like sequence; Octamer-binding protein 3-like; Octamer-binding transcription factor 3-like; OTF3C; OTF3P1; POU 5 domain protein; POU class 5 homeobox 1 pseudogene 1; POU class 5 homeobox 1B; POU domain transcription factor Oct-4; POU domain transcription factor OCT4-pg1; POU domain, class 5, transcription factor 1 pseudogene 1; POU5F1B; POU5F1P1; POU5F1P4; POU5FLC20; POU5FLC8; Putative POU domain, class 5, transcription factor 1B
Common Name POU5F1B
Gene Symbol POU5F1B
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VGPGSEVWGIPPCPPPYELCGGMAYCGPQVGVGLVPQGGLETSQPESEAGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.