Learn More
Indigo Biosciences Human PPAR Reporter Assays Panel All-inclusive Cell-Based Luciferase Reporter Assay Kit, Including PPARa, PPARd, PPARg, each in 1x32 96-well format
Catalog No. 502231760
Encompass_Preferred
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO's Human PPAR Panel (PPAR alpha, PPAR delta, and PPAR gamma) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each PPAR, optimized growth media, media for diluting test compounds, a control agonist for each PPAR, luciferase detection reagent, detailed protocol, and a protocol quick guide, and a cell culture-ready assay plate in strip-well format so that (if preferred) PPAR alpha, PPAR delta, and PPAR gamma assays may be performed at different times. INDIGO’s unique Cryomite™ process provides exceptional cell viability post-thaw and INDIGO’s strict QA process means consistent results lot to lot (product quality data available upon request). This kit allows researchers to achieve superior sensitivity, clear, and reproducible results.
This item is not returnable. View return policy
Catalog No. 50-223-1760 Supplier Indigo Biosciences Supplier No. IB0013132P
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO's Human PPAR Panel (PPAR alpha, PPAR delta, and PPAR gamma) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each PPAR, optimized growth media, media for diluting test compounds, a control agonist for each PPAR, luciferase detection reagent, detailed protocol, and a protocol quick guide, and a cell culture-ready assay plate in strip-well format so that (if preferred) PPAR alpha, PPAR delta, and PPAR gamma assays may be performed at different times. INDIGO’s unique Cryomite™ process provides exceptional cell viability post-thaw and INDIGO’s strict QA process means consistent results lot to lot (product quality data available upon request). This kit allows researchers to achieve superior sensitivity, clear, and reproducible results.