Learn More
Sino Biological Human PPARA Protein 50ug (Sino Biological)
Supplier:  Sino Biological 12080H07E50
Encompass_Preferred
A DNA sequence encoding the Human PPARA (Q07869) (Thr200-Tyr468) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3921