Learn More
Sino Biological Human PPCDC MAb 100ul (Sino Biological)
Supplier:  Sino Biological 14572MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PPC-DC / PPCDC (rh PPC-DC / PPCDC; Catalog#14572-H07E; AAH14409.1; Met1-Ser204). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1553