Learn More
Sino Biological Human PPCDC PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14572T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PPC-DC / PPCDC (rh PPC-DC / PPCDC; Catalog#14572-H07E; AAH14409.1; Met1-Ser204). PPC-DC / PPCDC specific IgG was purified by Human PPC-DC / PPCDC affinity chromatography.
Catalog No. 50-237-3549