Learn More
Sino Biological Human PPM1A MAb 100TS (Sino Biological)
Supplier:  Sino Biological 12984MM01P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PPM1A / PP2C-alpha (rh PPM1A / PP2C-alpha; Catalog#12984-H08E; P35813-1; Met1-Trp382) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-2681