Learn More
Sino Biological Anti-PPM1A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12984T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PPM1A (rh PPM1A; Catalog#12984-H08E; P35813-1; Met1-Trp382). PPM1A specific IgG was purified by Human PPM1A affinity chromatography.
Catalog No. 50-241-1121