Learn More
Sino Biological Anti-PPM1G Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11245RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PPM1G (rh PPM1G; Catalog#11245-H07E; O15355; Met 317-Asp 546). PPM1G specific IgG was purified by Human PPM1G affinity chromatography.
Catalog No. 50-241-1151