Learn More
Sino Biological Anti-PPP2R1A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 108140T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PPP2R1A, and purified by antigen affinity chromatography.
Catalog No. 50-241-2263