Learn More
Sino Biological Anti-PPP4C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105531T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PPP4C, and purified by antigen affinity chromatography.
Catalog No. 50-241-1500