Learn More
Bon Opus Biosciences Recombinant Human Palmitoyl-Protein Thioesterase 1/PPT1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CC641MG
Encompass_Preferred
Recombinant Human Palmitoyl-protein thioesterase 1 is produced by our Mammalian expression system and the target gene encoding Asp28-Gly306 is expressed with a 6His tag at the C-terminus. Accession# P50897. Formulation: Supplied as a 0.2 μm filtered solution of 20mM TrisHCl, 150mM NaCl, 10% Glycerol, pH 7.5 Bon Opus Cat# CC64
Catalog No. 50-217-2774