Learn More
Sino Biological Anti-PPT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14703T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PPT1 / Palmitoyl-protein thioesterase 1 (rh PPT1 / Palmitoyl-protein thioesterase 1; Catalog#14703-H08H; P50897-1; Met1-Gly306). PPT1 / Palmitoyl-protein thioesterase 1 specific IgG was purified by Human PPT1 / Palmitoyl-protein thioesterase 1 affinity chromatography.
Catalog No. 50-241-1095