Learn More
Sino Biological Human PRAC1 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 15425H28E50
Encompass_Preferred
A DNA sequence encoding the human PRAC (NP_115767.1) (Met1-Phe57) was expressed with a N-terminal polyhistide-tagged SUMO tag at the N-terminus.
Catalog No. 50-237-2371