Learn More
Sino Biological Human PRDX1 PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11194RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Peroxiredoxin 1 / PRDX1 (rh Peroxiredoxin 1 / PRDX1; Catalog#11194-H08E; NP_002565.1; Met 1-Lys 199). Peroxiredoxin 1 / PRDX1 specific IgG was purified by Human Peroxiredoxin 1 / PRDX1 affinity chromatography.
Catalog No. 50-237-1498