Learn More
Sino Biological Human PRDX6 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11257MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Peroxiredoxin 6 / PRDX6 (rh Peroxiredoxin 6 / PRDX6; Catalog#11257-H07E; P30041; Met 1-Pro 224). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3482