Learn More
Amsbio LLC Human Preadipocyte Growth Medium
Catalog No. 501632196
Encompass
Preadipocyte Growth Medium was designed to promote attachment, spreading and proliferation of preadipocytes in culture. This medium is fully supplemented with fetal bovine serum, growth factors and antibiotics. Preadipocyte Growth Medium is sterile filtered, bicarbonate buffered and is ready for use.
Catalog No. 50-163-2196 Supplier Amsbio LLC Supplier No. AMS.PM811500
Preadipocyte Growth Medium was designed to promote attachment, spreading and proliferation of preadipocytes in culture. This medium is fully supplemented with fetal bovine serum, growth factors and antibiotics. Preadipocyte Growth Medium is sterile filtered, bicarbonate buffered and is ready for use.