Learn More
Biomatik Corporation Human Prealbumin (PALB) CLIA Kit
Supplier:  Biomatik Corporation EKU09843
Encompass_Preferred
Human Prealbumin (PALB) CLIA Kit for detecting PALB in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 8.23-6,000pg/ml. Sensitivity: 3.16pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02766. Gene ID: 7276. Target Synonyms: TBPA; PA; PLB; ATTR; TTR; Transthyretin; Prealbumin Amyloidosis Type I; Transports Thyroxine and Retinol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8419