Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Pref-1 ELISA Kit

Catalog No. EH379RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH379RBX5 5 x 96 Tests
EH379RB 96 Tests
EH379RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH379RBX5 Supplier Invitrogen™ Supplier No. EH379RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human Pref-1 quantitates human Pref-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Pref-1.

The Delta-like 1 homolog (DLK1) is a transmembrane protein containing six epidermal growth factor repeats. It is involved in the differentiation of several cell types, including adipocytes and is also thought to be a tumor suppressor. The DLK1 gene is one of several imprinted genes located in a region on chromosome 14q32; certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). DLK1 is expressed from the paternal allele; a polymorphism within this gene has been associated with child and adolescent obesity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80370
Assay Range 0.4-100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Adipocyte differentiation inhibitor protein; AW742678; delta like non-canonical Notch ligand 1; Delta1; delta-like 1 homolog; delta-like 1 homolog (Drosophila); DLK; Dlk1; DLK-1; DlkI; FA1; Fetal antigen 1; Ly107; Peg9; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref1; pref-1; Protein delta homolog 1; SCP1; Scp-1; secredeltin; stromal cell derived protein 1; ZOGAdipocyte differentiation inhibitor protein; AW742678; delta like non-canonical Notch ligand 1; Delta1; delta-like 1 homolog; delta-like 1 homolog (Drosophila); DLK; Dlk1; DLK-1; DlkI; FA1; Fetal antigen 1; Ly107; Peg9; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref1; pref-1; Protein delta homolog 1; SCP1; Scp-1; secredeltin; stromal cell derived protein 1; ZOG;
Gene ID (Entrez) 8788
Gene Symbol DLK1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target DLK1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.