Learn More
Biomatik Corporation Human Prefoldin Subunit 1 (PFDN1) ELISA Kit
Supplier:  Biomatik Corporation EKU06750
Encompass_Preferred
Human Prefoldin Subunit 1 (PFDN1) ELISA Kit for detecting PFDN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60925. Gene ID: 5201. Target Synonyms: PDF; PFD1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8810