Learn More
American Research Products Inc Human pregnancy associated pla
Catalog No. 5000326454
Encompass
Human pregnancy associated plasma protein A PAPP-A ELISA Kit from CUSABIO Detection Range 1 7 ng/mL-50 ng/mL Sensitivity 0 4 ng/mL Sample serum plasma urine Method Sandwich
Catalog No. 50-003-26454 Supplier American Research Products Inc Supplier No. CSBE08220H
Add to Cart
Add to Cart
Human pregnancy associated plasma protein A PAPP-A ELISA Kit from CUSABIO Detection Range 1 7 ng/mL-50 ng/mL Sensitivity 0 4 ng/mL Sample serum plasma urine Method Sandwich