Learn More
American Research Products Inc Human Pregnancy specific beta
Catalog No. 5000327166
Encompass
Human Pregnancy-specific beta-1-glycoprotein 9 (PSG9) ELISA kit from CUSABIO Detection Range 5 g/mL-150 g/mL Sensitivity 1 25 g/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27166 Supplier American Research Products Inc Supplier No. CSBEL018859HU
Add to Cart
Add to Cart
Human Pregnancy-specific beta-1-glycoprotein 9 (PSG9) ELISA kit from CUSABIO Detection Range 5 g/mL-150 g/mL Sensitivity 1 25 g/mL Sample serum plasma Method Sandwich