Learn More
Biomatik Corporation Human Pregnancy Specific Beta-1-Glycoprotein 2 (PSG2) ELISA Kit
Supplier:  Biomatik Corporation EKU06753
Encompass_Preferred
Human Pregnancy Specific Beta-1-Glycoprotein 2 (PSG2) ELISA Kit for detecting PSG2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11465. Gene ID: 5670. Target Synonyms: CEA; PSG1; PSBG2; PSGGB; Carcinoembryonic Antigen SG8; Pregnancy-specific glycoprotein 2; Pregnancy-specific beta-1 glycoprotein E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6251