Learn More
Enzyme Research Laboratories Inc Human Prekallikrein 1.0mg Vial
Catalog No. NC9937651
Encompass
Our plama purified Prekallikrein is a single chain gamma globulin glycoprotein that participates in the early phase of contact activation, kinin formation and fibrinolysis. Purity is determined by SDS-PAGE.
Catalog No. NC9937651 Supplier Enzyme Research Laboratories Inc Supplier No. HPK 1302
Our plama purified Prekallikrein is a single chain gamma globulin glycoprotein that participates in the early phase of contact activation, kinin formation and fibrinolysis. Purity is determined by SDS-PAGE.