Learn More
Biomatik Corporation Human Preprovasopressin (VP) ELISA Kit
Supplier:  Biomatik Corporation EKU11354
Encompass_Preferred
Human Preprovasopressin (VP) ELISA Kit for detecting VP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01185. Gene ID: 551. Target Synonyms: AVP-NPII; ARVP; AVP; Vasopressin-Neurophysin 2-Copeptin; Arginine-vasopressin; Neurophysin 2; Copeptin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6863