Learn More
Raybiotech Inc Human Presenilin 1 ELISA Kit
Supplier:  Raybiotech Inc ELHPSEN11
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 10.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human B7-2 (CD86), BAFF R, Calcitonin, Calsyntenin-1, Cathepsin E, cIAP-2 (HIAP-1), Coagulation Factor VII, Complement MASP3, Endocan, EphA2, EphB4, Ephrin-A4, FGF-23, FGF-5, Flt-3 (Flk-2), GLP-1 (7-37 & 7-36-NH2), Glypican 2, GM-CSF R alpha, GP73 (GOLM1), HTRA2 (Omi), IL-20 R alpha, IL-4 R alpha, JAM-C, LH, Matrilin-3, Meprin alpha (MEP1A), MSP R (Ron), N-Cadherin, Neprilysin-2 (MMEL1), NKp44, PAPP-A, PTH, Pepsinogen II (PGC), PYY, SOX2, TFPI-2, TRACP (ACP5),TFF3, Ubiquitin+1.
Catalog No. 50-194-9668