Learn More
Biomatik Corporation Human Presenilin Associated Metalloprotease (PAMP) ELISA Kit
Supplier:  Biomatik Corporation EKU09318
Encompass_Preferred
Human Presenilin Associated Metalloprotease (PAMP) ELISA Kit for detecting PAMP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96TA2. Target Synonyms: FTSH; MEG4; YME1L1; YME1L; ATP-dependent metalloprotease FtsH1; YME1 Like Protein 1; ATP-dependent zinc metalloprotease YME1L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6577