Learn More
Sino Biological Recombinant Human PRG2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16787H08H100
Encompass_Preferred
A DNA sequence encoding the Human PRG2 (P13727) (Met1-Tyr222) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4016