Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRG4 Partial ORF (NP_005798, 1305 a.a. - 1404 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098550 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-085 25 μg
89-958-084 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-085 Supplier Abnova™ Supplier No. H00010216Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a large proteoglycan specifically synthesized by chondrocytes located at the surface of articular cartilage, and also by some synovial lining cells. This protein contains both chondroitin sulfate and keratan sulfate glycosaminoglycans. It functions as a boundary lubricant at the cartilage surface and contributes to the elastic absorption and energy dissipation of synovial fluid. Mutations in this gene result in camptodactyly-arthropathy-coxa vara-pericarditis syndrome. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: ERAIGPSQTHTIRIQYSPARLAYQDKGVLHNEVKVSILWRGLPNVVTSAISLPNIRKPDGYDYYAFSKDQYYNIDVPSRTARAITTRSGQTLSKVWYNCP

Specifications

Accession Number NP_005798
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10216
Molecular Weight (g/mol) 36.74kDa
Name Human PRG4 Partial ORF (NP_005798, 1305 a.a. - 1404 a.a.) Recombinant Protein with GST-tag at N-terminal
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CACP, FLJ32635, HAPO, JCAP, MSF, SZP, bG174L6.2
Common Name PRG4
Gene Symbol PRG4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.