Learn More
BMEM BIO Human Primed Pluripotent Media
Catalog No. NC3923311
Encompass
BMEM Human Primed Pluripotent is a highly optimized, chemically defined, animal origin-free (AOF) basal medium and supplement combination for the maintenance and expansion of human induced pluripotent stem cells (hiPSCs) in adherent culture.•Optimized Formulation: Features a new Primed Supplement designed to ease the transition of hiPSCs from legacy media (e.g., mTeSR1™), resolving a key challenge of chemically defined systems.•Enhanced Performance: Promotes increased cell proliferation and upregulation of core pluripotency factors.•Protocol Friendly: Supports weekend-free and no-change feeding schedules (from original full text).•Broad Compatibility: Works with diverse matrices (Matrigel, Geltrex™, Vitronectin) and passaging methods (TrypLE™, ReLeSR™, Versene).•Quality & Stability: Supplement is optimized for stability (allowing room temperature shipment). Each lot is performance tested on hiPSCs and confirmed sterile.
Catalog No. NC3923311 Supplier BMEM BIO Supplier No. 1001
BMEM Human Primed Pluripotent is a highly optimized, chemically defined, animal origin-free (AOF) basal medium and supplement combination for the maintenance and expansion of human induced pluripotent stem cells (hiPSCs) in adherent culture.•Optimized Formulation: Features a new Primed Supplement designed to ease the transition of hiPSCs from legacy media (e.g., mTeSR1™), resolving a key challenge of chemically defined systems.•Enhanced Performance: Promotes increased cell proliferation and upregulation of core pluripotency factors.•Protocol Friendly: Supports weekend-free and no-change feeding schedules (from original full text).•Broad Compatibility: Works with diverse matrices (Matrigel, Geltrex™, Vitronectin) and passaging methods (TrypLE™, ReLeSR™, Versene).•Quality & Stability: Supplement is optimized for stability (allowing room temperature shipment). Each lot is performance tested on hiPSCs and confirmed sterile.