Learn More
Biomatik Corporation Human Prion Protein (PRNP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11967
Encompass_Preferred
Human Prion Protein (PRNP) Mini sample ELISA Kit for detecting PRNP in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04156. Target Synonyms: CD230; GSS; ASCR; CJD; PRIP; PrP; PrP2730; PrPc; Major prion protein; Creutzfeldt-Jakob Disease; Gerstmann-Strausler-Scheinker Syndrome; Fatal Familial Insomnia. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5773