Learn More
Abcam Human Prion protein PrP ELISA Kit, 1 x 96 Tests
Catalog No. 502651154
Encompass_Preferred
Human Prion protein PrP ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Prion protein PrP with a sensitivity of 19.817 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-1154 Supplier Abcam Supplier No. AB3150451X96TESTS
Human Prion protein PrP ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Prion protein PrP with a sensitivity of 19.817 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement