Learn More
Sino Biological Anti-AMPK alpha 1/PRKAA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102146T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human AMPK alpha 1/PRKAA1, and purified by antigen affinity chromatography.
Catalog No. 50-240-9006