Learn More
BIOHIPPO Human Pro-ANP Microsample
SDP
Catalog No. 5000315216
Encompass
Human Pro-ANP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Pro-ANP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human Pro-ANP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human Pro-ANP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human Pro-ANP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15216 Supplier BIOHIPPO Supplier No. BHE15210527
Human Pro-ANP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Pro-ANP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human Pro-ANP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human Pro-ANP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human Pro-ANP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color