Learn More
Raybiotech Inc Human/Mouse/Rat Pro-Neuropeptide Y/CPON EIA Kit.
Supplier:  Raybiotech Inc EIAPRONPY1
Encompass_Preferred
Optimized for serum and cell culture supernatants. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8837