Learn More
Biomatik Corporation Human Profilin 2 (PFN2) ELISA Kit
Supplier:  Biomatik Corporation EKU11362
Encompass_Preferred
Human Profilin 2 (PFN2) ELISA Kit for detecting PFN2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.137ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35080. Gene ID: 5217. Target Synonyms: PFL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8158