Learn More
American Research Products Inc Human Programmed cell death 6
Supplier:  American Research Products Inc CSBEL017673HU
Encompass
Human Programmed cell death 6-interacting protein (PDCD6IP) ELISA kit from CUSABIO Detection Range 47 pg/mL-3000 pg/mL Sensitivity 11 7 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28039