Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Prolactin R (NP_000940) VersaClone cDNA

Catalog No. RDC2562
Change view
Click to view available options
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
RDC2562 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2562 Supplier R&D Systems™ Supplier No. RDC2562
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • PRLR prolactin receptor [ Homo sapiens (human) ]
  • Also known as: HPRL; MFAB; hPRLrI; RI-PRLR
  • The Prolactin R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • PRLR is a receptor for the anterior pituitary hormone, prolactin, and belongs to the type I cytokine receptor family.
  • Prolactin-dependent signaling occurs as the result of ligandinduced dimerization of the prolactin receptor.
  • Several alternatively spliced transcript variants encoding different membrane-bound and soluble isoforms have been described for PRLR, which may function to modulate the endocrine and autocrine effects of prolactin in normal tissue and cancer.
  • For reasearch use only. Not for use in humans.

Specifications

Gene hPRLR
Accession Number NP_000940
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1882 bp
Gene ID (Entrez) 5618
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.