Learn More
Sino Biological Human Prolactin Receptor Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785436
Encompass_Preferred
The human PRLR / Prolactin Receptor ELISA Pair Set is for the quantitative determination of human PRLR.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for PRLR / Prolactin Receptor coated on a 96-well plate. Standards and samples are added to the wells, and any PRLR / Prolactin Receptor present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-PRLR monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of PRLR / Prolactin Receptor present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85436 Supplier Sino Biological Supplier No. SEK1027815
The human PRLR / Prolactin Receptor ELISA Pair Set is for the quantitative determination of human PRLR.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for PRLR / Prolactin Receptor coated on a 96-well plate. Standards and samples are added to the wells, and any PRLR / Prolactin Receptor present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-PRLR monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of PRLR / Prolactin Receptor present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.