Learn More
Biomatik Corporation Human Proliferation Associated Protein 2G4 (PA2G4) ELISA Kit
Supplier:  Biomatik Corporation EKU06828
Encompass_Preferred
Human Proliferation Associated Protein 2G4 (PA2G4) ELISA Kit for detecting PA2G4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UQ80. Gene ID: 5036. Target Synonyms: EBP1; HG4-1; p38-2G4; Cell cycle protein p38-2G4 homolog; ErbB3-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5623