Learn More
Biomatik Corporation Human Prolyl-4-Hydroxylase Alpha Polypeptide II (P4Ha2) ELISA Kit
Supplier:  Biomatik Corporation EKU06833
Encompass_Preferred
Human Prolyl-4-Hydroxylase Alpha Polypeptide II (P4Ha2) ELISA Kit for detecting P4Ha2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15460. Gene ID: 8974. Target Synonyms: Procollagen-Proline,2-Oxoglutarate 4-Dioxygenase (Proline 4-Hydroxylase) ,Alpha Polypeptide II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8805