Learn More
Biomatik Corporation Human Prolyl-4-Hydroxylase Alpha Polypeptide III (P4Ha3) ELISA Kit
Supplier:  Biomatik Corporation EKU06834
Encompass_Preferred
Human Prolyl-4-Hydroxylase Alpha Polypeptide III (P4Ha3) ELISA Kit for detecting P4Ha3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7Z4N8. Gene ID: 283208. Target Synonyms: Procollagen-Proline,2-Oxoglutarate 4-Dioxygenase (Proline 4-Hydroxylase) ,Alpha Polypeptide III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7342