Learn More
Biomatik Corporation Human Promyelocytic Leukemia Protein (PML) ELISA Kit
Supplier:  Biomatik Corporation EKU06838
Encompass_Preferred
Human Promyelocytic Leukemia Protein (PML) ELISA Kit for detecting PML in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29590. Gene ID: 5371. Target Synonyms: MYL; RNF71; TRIM19; Probable Transcription Factor PML; Ring Finger Protein 71; Tripartite motif-containing protein 19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4689