Learn More
Raybiotech Inc Human Properdin ELISA Kit
Supplier:  Raybiotech Inc ELHPROPE1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 42 pg/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Properdin.
Catalog No. 50-194-9663