Learn More
Biomatik Corporation Human Prostaglandin E Receptor 2 (EP2) ELISA Kit
Supplier:  Biomatik Corporation EKU08334
Encompass_Preferred
Human Prostaglandin E Receptor 2 (EP2) ELISA Kit for detecting EP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.071ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43116. Gene ID: 5732. Target Synonyms: PTGER2; Prostaglandin E2 Receptor (Subtype EP2) ,53kD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4771